Learn More
Medchemexpress LLC 8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY | 250734-47-5 | MFCD29905036 | 98.5% | 450.07 g·mol⁻¹ | C19H18BF2IN2 | 250 MG
SDP
Catalog No. 5000282070
Encompass_Preferred
8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is a halogenated BODIPY fluorescent dye designed for receptor labeling and cellular imaging. It exhibits strong fluorescence typical of BODIPY scaffolds and is used as a probe in studies involving GFP-labeled muscarinic M1 receptors and related fluorescence applications.
Catalog No. 50-002-82070 Supplier Medchemexpress LLC Supplier No. HYW248119250MG
8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is a halogenated BODIPY fluorescent dye designed for receptor labeling and cellular imaging. It exhibits strong fluorescence typical of BODIPY scaffolds and is used as a probe in studies involving GFP-labeled muscarinic M1 receptors and related fluorescence applications.