Learn More
Medchemexpress LLC 8-(4-(trifluoromethyl)phenyl)-N-(1,1,1-trifluoropropan-2-yl)-dihydro-naphthyridine-7-carboxamide | 1470018-52-0 | 99.9% | 417.35 g/mol | C19H17F6N3O | 5 MG
SDP
Catalog No. 5000243209
Encompass_Preferred
AMG2850 is a potent, orally bioavailable, and selective antagonist of the TRPM8 ion channel developed for preclinical research. It blocks menthol- and cold-induced TRPM8 activation in cellular assays and has been characterized in pharmacology studies assessing sensory signaling and pain mechanisms. Supplied as a solid research chemical for laboratory use only.
  • Potent, selective TRPM8 antagonist.
  • Orally bioavailable in preclinical models.
  • Inhibits menthol- and cold-induced TRPM8 activation in vitro.
  • Supplied as a solid with high purity suitable for research.
  • Useful for studies of sensory signaling and pain mechanisms.
Catalog No. 50-002-43209 Supplier Medchemexpress LLC Supplier No. HY1040595MG
Add to Cart
Add to Cart
AMG2850 is a potent, orally bioavailable, and selective antagonist of the TRPM8 ion channel developed for preclinical research. It blocks menthol- and cold-induced TRPM8 activation in cellular assays and has been characterized in pharmacology studies assessing sensory signaling and pain mechanisms. Supplied as a solid research chemical for laboratory use only.
  • Potent, selective TRPM8 antagonist.
  • Orally bioavailable in preclinical models.
  • Inhibits menthol- and cold-induced TRPM8 activation in vitro.
  • Supplied as a solid with high purity suitable for research.
  • Useful for studies of sensory signaling and pain mechanisms.