Learn More
Cayman Chemical 8 4azIdophenacylthIo NAD 900ug
SDP
Catalog No. 502918374
Encompass_Preferred
A clickable form of NAD+
Catalog No. 50-291-8374 Supplier Cayman Chemical Supplier No. 38833900 UG
A clickable form of NAD+