Learn More
Cayman Chemical 8(4'-bromophenyl)-1,3,5,7-tetramethyl-BODIPY, Purity: ≥95%, CAS: 850534-66-6, 100 mg
SDP
Catalog No. 502942417
Encompass_Preferred
An intermediate in the synthesis of BODIPY-containing dyes; has been used as a precursor in the synthesis of BODIPY-containing photosensitizer dyes with antibacterial activity
Catalog No. 50-294-2417 Supplier Cayman Chemical Supplier No. 40123100 MG
An intermediate in the synthesis of BODIPY-containing dyes; has been used as a precursor in the synthesis of BODIPY-containing photosensitizer dyes with antibacterial activity