Learn More
Quality Lab Accessories 8.5 COIL SPIRAL SS SINKER
Catalog No. NC9766469
Encompass
Spiral Coated Capsule Sinker, 316 SS, .885" (22.5mm) L x .575" (14.6mm) W capacity, 8.5 coils
Catalog No. NC9766469 Supplier Quality Lab Accessories Supplier No. CAPWHTXS
Spiral Coated Capsule Sinker, 316 SS, .885" (22.5mm) L x .575" (14.6mm) W capacity, 8.5 coils