Learn More
Medchemexpress LLC 8-[5-(difluoromethyl)pyridin-3-yl]-1-(2,3,6,7-tetrahydrooxepin-4-yl)triazolo[4,3-a]quinoxaline | 2763602-59-9 | 99.4% | C21H17F2N5O | 10MG
SDP
Supplier:  Medchemexpress LLC HY13229310MG
Encompass_Preferred
BAY-8400 is an orally active, potent, and selective DNA-dependent protein kinase (DNA-PK) inhibitor (IC50 = 81 nM) used in oncology research. It has shown antiproliferative effects in multiple cancer cell lines and oral bioavailability in vivo; supplier data report high purity, defined storage conditions, and good solubility in DMSO, making it suitable for in vitro and in vivo research applications.
  • Potent DNA-PK inhibition (IC50 81 nM).
  • Orally active with reported in vivo bioavailability.
  • Antiproliferative activity in multiple cancer cell lines.
  • High reported purity for research use.
  • Good DMSO solubility for in vitro assays.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-60771