Learn More
ELCOMETER INC 8.5 FL OZ PURE WATER- GRADE 3
SDP
Catalog No. NC2283506
Encompass
BOTTLE OF PURE WATER FOR ELCOMETER MODEL 130 8.5 floz / 250ml. Grade 3. Meets the ISO 3696 (1987) and BS3978(1967) standards for Water for Lab Use. Includes custom cap forattachment of Elcometer T13024091 needle-less syringes.
This item is not returnable. View return policy
Catalog No. NC2283506 Supplier ELCOMETER INC Supplier No. T99911344
BOTTLE OF PURE WATER FOR ELCOMETER MODEL 130 8.5 floz / 250ml. Grade 3. Meets the ISO 3696 (1987) and BS3978(1967) standards for Water for Lab Use. Includes custom cap forattachment of Elcometer T13024091 needle-less syringes.