Learn More
Cayman Chemical 8 6AmInohexylthIogunosIn 700ug
SDP
Catalog No. 502918875
Encompass_Preferred
An aminohexylthiol-substituted derivative of GTP; has been used in the synthesis of biotinylated derivatives of GTP
Catalog No. 50-291-8875 Supplier Cayman Chemical Supplier No. 38811700 UG
An aminohexylthiol-substituted derivative of GTP; has been used in the synthesis of biotinylated derivatives of GTP