Learn More
Cayman Chemical 8 9-EET methyl ester 100UG
SDP
Catalog No. 502894933
Encompass_Preferred
An esterified version of (±)8(9)-EET; more stable for long-term storage and can be readily hydrolyzed to the free acid just before use
Catalog No. 50-289-4933 Supplier Cayman Chemical Supplier No. 50350100 UG
An esterified version of (±)8(9)-EET; more stable for long-term storage and can be readily hydrolyzed to the free acid just before use