Learn More
Cayman Chemical 8 9DiHETd11 GC MS Standards
SDP
Catalog No. 502889051
Encompass_Preferred
An internal standard for the quantification of 8(9)-DiHET by GC- or LC-MS
Catalog No. 50-288-9051 Supplier Cayman Chemical Supplier No. 10009998100 UG
An internal standard for the quantification of 8(9)-DiHET by GC- or LC-MS