Learn More
Cayman Chemical 8 9DiHETd11 GC MS Standards
SDP
Supplier:  Cayman Chemical 1000999850 UG
Encompass_Preferred
An internal standard for the quantification of 8(9)-DiHET by GC- or LC-MS
Catalog No. 50-288-9122