Learn More
Cayman Chemical 8 9EETd11 GC MS Standards
SDP
Supplier:  Cayman Chemical 1000953225 UG
Encompass_Preferred
An internal standard for the quantification of (±)8(9)-EET by GC- or LC-MS
Catalog No. 50-288-9058