Learn More
Cayman Chemical 8 9EETd11 GC MS Standards
SDP
Catalog No. 502888002
Encompass_Preferred
An internal standard for the quantification of (±)8(9)-EET by GC- or LC-MS
Catalog No. 50-288-8002 Supplier Cayman Chemical Supplier No. 1000953210 UG
An internal standard for the quantification of (±)8(9)-EET by GC- or LC-MS