Learn More
Cayman Chemical 8 9EETd11 methyl ester 100ug
SDP
Catalog No. 502988135
Encompass_Preferred
An internal standard for the quantification of 8(9)-EET methyl ester by GC- or LC-MS
Catalog No. 50-298-8135 Supplier Cayman Chemical Supplier No. 29400100 UG
An internal standard for the quantification of 8(9)-EET methyl ester by GC- or LC-MS