Learn More
Cayman Chemical 8 9EpETE Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1047050 UG
Encompass_Preferred
An epoxygenase pathway product produced from EPA by CYP450 both in vitro and in vivo
Catalog No. 50-288-9608