Learn More
Abcam 8-Aceto x ypyrene-1,3,6-trisulfonic acid trisodium salt, Fluorogenic esterase substrate, 1G
Catalog No. 502705229
Encompass_Preferred
MW 566.4 Da, Purity >98%. Highly water soluble substrate for fluorometric assay of esterases at longer wavelengths.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5229 Supplier Abcam Supplier No. AB1454001G
MW 566.4 Da, Purity >98%. Highly water soluble substrate for fluorometric assay of esterases at longer wavelengths.
The product is subject to the following: Abcam Restricted Use Statement