Learn More
Covaris Inc 8 AFA-TUBE TPX STRIP
SDP
Catalog No. NC2002573
Encompass
8 AFA-TUBE TPX STRIPFragment Size: 175 bp to 1.5 kbSample Volume: 10 to 50 μlApplication: FFPE Extraction, Chromatin Shearing, DNA/RNA Shearing, Cell Lysis, Proteomics, Whole Blood Extraction.COVARIS INC mfg# 520292
Catalog No. NC2002573 Supplier Covaris Inc Supplier No. 520292
8 AFA-TUBE TPX STRIPFragment Size: 175 bp to 1.5 kbSample Volume: 10 to 50 μlApplication: FFPE Extraction, Chromatin Shearing, DNA/RNA Shearing, Cell Lysis, Proteomics, Whole Blood Extraction.COVARIS INC mfg# 520292