Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Amino-2-methylquinoline 98.0+%, TCI America™
SDP

Catalog No. A04165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04165G 5 g
A04161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04165G Supplier TCI America Supplier No. A04165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18978-78-4
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00023998
InChI Key JHIAOWGCGNMQKA-UHFFFAOYSA-N
Synonym 8-Aminoquinaldine
PubChem CID 140457
IUPAC Name 2-methylquinolin-8-amine
SMILES CC1=CC=C2C=CC=C(N)C2=N1

Specifications

Melting Point 56°C
Color Yellow
Quantity 5 g
Formula Weight 158.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Amino-2-methylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.