Learn More
Cayman Chemical (±)8-Amino-7-Oxononanoic Acid (hydrochloride), Purity: ≥95%, CAS: 21286-96-4, 10 mg
SDP
Catalog No. 502944970
Encompass_Preferred
A racemic mixture of 8(R)-KAPA and 8(S)-KAPA; biosynthesized as the 8(S) form but racemizes in culture media in a pH-dependent manner, with a higher racemization rate under very low (pH E. coli and M. tuberculosis DAPA AT, which leads to accumulation of 8(R)-KAPA in M. tuberculosis cultures after exogenous application of racemic KAPA
Catalog No. 50-294-4970 Supplier Cayman Chemical Supplier No. 4097310 MG
A racemic mixture of 8(R)-KAPA and 8(S)-KAPA; biosynthesized as the 8(S) form but racemizes in culture media in a pH-dependent manner, with a higher racemization rate under very low (pH E. coli and M. tuberculosis DAPA AT, which leads to accumulation of 8(R)-KAPA in M. tuberculosis cultures after exogenous application of racemic KAPA