Learn More
Medchemexpress LLC 8-Aminoguanine | 28128-41-8 | 97.19% | 166.14 | 100 MG
SDP
Catalog No. 5000347799
Encompass_Preferred
8-Aminoguanine is an endogenous purine that inhibits purine nucleoside phosphorylase (PNPase) and induces diuresis, natriuresis, and glucosuria. This product is intended for research use only.
  • Inhibits purine nucleoside phosphorylase (PNPase)
  • Induces diuresis, natriuresis, and glucosuria
  • Exhibits cellular effects on MOLT-4 cell line
Catalog No. 50-003-47799 Supplier Medchemexpress LLC Supplier No. HY139037100MG
8-Aminoguanine is an endogenous purine that inhibits purine nucleoside phosphorylase (PNPase) and induces diuresis, natriuresis, and glucosuria. This product is intended for research use only.
  • Inhibits purine nucleoside phosphorylase (PNPase)
  • Induces diuresis, natriuresis, and glucosuria
  • Exhibits cellular effects on MOLT-4 cell line