Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-aminooctanoic acid | 1002-57-9 | MFCD00008245 | 98.0% | 159.23 | C8H17NO2 | 100 G
SDP

Catalog No. 5000300855
Encompass_Preferred

8-aminooctanoic acid is an omega-amino fatty acid (octanoic acid bearing a terminal amino group) used as a biochemical reagent and research compound. It is supplied as a powder for laboratory use and is commonly used in biochemical assays and analytical studies.

  • Purity 98.0%.
  • Molecular formula C8H17NO2.
  • Molecular weight 159.23 g/mol.
  • Supplied as a powder in multiple package sizes (5 g-100 g and bulk inquiry).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.
  • Function: omega-amino fatty acid used in biochemical research and assays.

Catalog No. 50-003-00855 Supplier Medchemexpress LLC Supplier No. HYW018678100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-aminooctanoic acid is an omega-amino fatty acid (octanoic acid bearing a terminal amino group) used as a biochemical reagent and research compound. It is supplied as a powder for laboratory use and is commonly used in biochemical assays and analytical studies.

  • Purity 98.0%.
  • Molecular formula C8H17NO2.
  • Molecular weight 159.23 g/mol.
  • Supplied as a powder in multiple package sizes (5 g-100 g and bulk inquiry).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.
  • Function: omega-amino fatty acid used in biochemical research and assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.