Learn More
Medchemexpress LLC 8-Aminopyrene-1,3,6-trisulfonic acid trisodium | 196504-57-1 | 99.75% | 25 MG
SDP
Catalog No. 5000412646
Encompass_Preferred
8-Aminopyrene-1,3,6-trisulfonic acid trisodium (APTS) is an anionic fluorescent dye with excitation/emission at 425/503 nm in pH 7.4 PBS. Its fluorescence intensity remains stable at pH 4-10. It can be used for glucose sensing and retains its fluorescence properties when immobilized in a hydrogel by copolymerization.
  • Anionic fluorescent dye (excitation/emission: 425/503 nm in pH 7.4 PBS)
  • Fluorescence intensity stable at pH 4-10
  • Retains fluorescence properties when immobilized in a hydrogel by copolymerization
  • Can be used for glucose sensing
Catalog No. 50-004-12646 Supplier Medchemexpress LLC Supplier No. 25MGHYD0031
8-Aminopyrene-1,3,6-trisulfonic acid trisodium (APTS) is an anionic fluorescent dye with excitation/emission at 425/503 nm in pH 7.4 PBS. Its fluorescence intensity remains stable at pH 4-10. It can be used for glucose sensing and retains its fluorescence properties when immobilized in a hydrogel by copolymerization.
  • Anionic fluorescent dye (excitation/emission: 425/503 nm in pH 7.4 PBS)
  • Fluorescence intensity stable at pH 4-10
  • Retains fluorescence properties when immobilized in a hydrogel by copolymerization
  • Can be used for glucose sensing