Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-Aminoquinoline | 578-66-5 | 144.17 | 10 G
SDP

Catalog No. 5000460246
Encompass_Preferred

8-Aminoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.

  • Biochemical reagent for life science research
  • Organic compound for life science research
  • For research use only
  • Not for sale to patients
  • Solid appearance, brown to black color
  • Molecular formula: C9H8N2
  • Store solid at 4°C, protect from light
  • Store in solvent at -80°C for 6 months; -20°C for 1 month (protect from light)
  • Soluble in DMSO: 100 mg/mL

Catalog No. 50-004-60246 Supplier Medchemexpress LLC Supplier No. HYW00731710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Aminoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.

  • Biochemical reagent for life science research
  • Organic compound for life science research
  • For research use only
  • Not for sale to patients
  • Solid appearance, brown to black color
  • Molecular formula: C9H8N2
  • Store solid at 4°C, protect from light
  • Store in solvent at -80°C for 6 months; -20°C for 1 month (protect from light)
  • Soluble in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.