Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-Aminoquinoline | 578-66-5 | 6809 | 144.17 | 500 G
SDP

Catalog No. 5000460298
Encompass_Preferred

8-Aminoquinoline is a biochemical reagent suitable for use as a biological material or organic compound in life science related research.

  • Solid appearance
  • Brown to black color
  • Melting point 62-65 °C
  • Boiling point 174 °C (26 mmHg)
  • Air sensitive
  • Shelf life of 5 years

Catalog No. 50-004-60298 Supplier Medchemexpress LLC Supplier No. HYW007317500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Aminoquinoline is a biochemical reagent suitable for use as a biological material or organic compound in life science related research.

  • Solid appearance
  • Brown to black color
  • Melting point 62-65 °C
  • Boiling point 174 °C (26 mmHg)
  • Air sensitive
  • Shelf life of 5 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.