Learn More
Medchemexpress LLC 8-Aminoquinoline | 578-66-5 | MFCD00006809 | 144.17 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007317
Encompass_Preferred
8-Aminoquinoline is a biochemical reagent and organic compound utilized in life science research. It exhibits moderate to weak fluorescence in low dielectric media and has applications in the preparation of base-stabilized terminal borylene complexes of osmium and the spectrophotometric determination of bivalent palladium.
  • Biochemical reagent for life science research
  • Organic compound
  • Moderate to weak fluorescence in low dielectric media
  • Used in base-stabilized terminal borylene complex preparation
  • Used in spectrophotometric determination of bivalent palladium
Catalog No. 50-004-49988