Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Anilino-1-naphthalene sulfonic acid, MP Biomedicals™

Catalog No. 0210563301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-105-63301 1 g
02-105-63310 10 g
02-105-63325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-105-63301 Supplier MP Biomedicals, Inc Supplier No. 0210563301
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82-76-8
Molecular Formula C16H13NO3S
Molecular Weight (g/mol) 299.344
InChI Key FWEOQOXTVHGIFQ-UHFFFAOYSA-N
Synonym 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid
PubChem CID 1369
ChEBI CHEBI:39708
IUPAC Name 8-anilinonaphthalene-1-sulfonic acid
SMILES C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O

Specifications

Color Gray to Gray-Green
Assay Percent Range 0.983
Quantity 1 g
Formula Weight 299.35
Physical Form Crystalline Powder
Chemical Name or Material 8-Anilino-1-naphthalenesulfonic acid, For HPLC analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.