Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Anilino-1-naphthalene-sulfonic Acid, Sodium Salt, MP Biomedicals

Catalog No. ICN209041 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209041 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209041 Supplier MP Biomedicals, Inc Supplier No. 0520904101

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1445-19-8
Molecular Formula C16H12NNaO3S
Molecular Weight (g/mol) 321.33
MDL Number MFCD00054279
InChI Key VUSNMYYVKMGHEH-UHFFFAOYSA-M
Synonym ans-na, sodium 8-anilinonaphthalene-1-sulphonate, 1-naphthalenesulfonic acid, 8-phenylamino-, monosodium salt, 1,8-phenyl acid sodium salt, sodium 8-anilino-1-naphthalenesulfonate, 1-naphthalenesulfonic acid, 8-phenylamino-, sodium salt 1:1, 82-76-8 parent, sodium 8-phenylamino naphthalene-1-sulfonate, 1-naphthalenesulfonic acid, 8-anilino-, monosodium salt, sodium 8-anilinonaphthalene-1-sulfonate
PubChem CID 23670839
IUPAC Name sodium 8-(phenylamino)naphthalene-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1

Specifications

Quantity 1 g
Formula Weight 321.32
Chemical Name or Material 8-Anilino-1-naphthalene-sulfonic Acid, Na Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.