Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Anilino-1-Naphthalenesulfonic Acid Hemimagnesium Salt Hydrate, Sigma-Aldrich

Catalog No. 01258952 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-952 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-952 Supplier MilliporeSigma Supelco Supplier No. 1041925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

suitable for fluorescence, ≥95.0% (T)

ANSA (8-Anilino-1-naphthalenesulfonic acid) is a hydrophobic polarity sensitive fluorescent dye useful as a site probe to detect conformational changes in cell and micelle membranes and molecules such as proteins. ANSA is also used as a fluorescent probe for the estimation of the critical micelle concentration (CMC) of surfactants.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18108-68-4

Specifications

Quantity 25 g
Product Type Fluorescence Reagent
Molecular Weight (g/mol) 310.49 g/mol
Purity ≥95%
Label Type Other Labels or Dyes
Color Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.