Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Anilino-1-Naphthalenesulfonic Acid, Spectrum™ Chemical
SDP

Catalog No. 18620020 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
5 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-020 5 kg
18-620-015 5 g
18-620-016 25 g
18-620-017 100 g
18-620-018 500 g
18-620-019 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-620-020 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A12975KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82-76-8

Specifications

Packaging Drum
Quantity 5 kg
Percent Purity 95%
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.