Learn More
Medchemexpress LLC 8-aza-2'-deoxyguanosine | 4546-73-0 | 99.9% | 268.23 g/mol | C9H12N6O4 | 10 MG
SDP
Catalog No. 5000285952
Encompass_Preferred
8-Aza-2'-deoxyguanosine is a purine nucleoside analog used in research for its antitumor properties. It acts by interfering with DNA synthesis and promoting apoptosis, making it useful for studies of nucleoside analog mechanisms and lymphoid malignancies.
  • Purine nucleoside analog with broad antitumor activity.
  • Inhibits DNA synthesis and induces apoptosis.
  • High chemical purity for reliable experimental results.
  • White to off-white solid, convenient for handling.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-85952 Supplier Medchemexpress LLC Supplier No. HY15447010MG
8-Aza-2'-deoxyguanosine is a purine nucleoside analog used in research for its antitumor properties. It acts by interfering with DNA synthesis and promoting apoptosis, making it useful for studies of nucleoside analog mechanisms and lymphoid malignancies.
  • Purine nucleoside analog with broad antitumor activity.
  • Inhibits DNA synthesis and induces apoptosis.
  • High chemical purity for reliable experimental results.
  • White to off-white solid, convenient for handling.
  • Stable under recommended storage conditions for long-term use.