Learn More
Apexbio Technology LLC 8-Aza-ATP 50ul (100 mM)
SDP
Catalog No. 5000064178
Encompass_Preferred
8-Aza-ATP is a structural analog of ATP characterized by the substitution of the carbon atom at the 8-position of the purine ring with nitrogen resulting in an 8-azapurine nucleotide moiety Due to this structural alteration 8-Aza-ATP competitively interacts with ATP-binding sites on enzymes thereby modulating their catalytic activities This compound serves as a tool in biochemical research to elucidate enzyme-substrate recognition nucleotide binding specificity and catalytic mechanisms of ATP-dependent enzymes Additionally it is utilized in the study of cellular energy metabolism signal transduction pathways and nucleotide-mediated regulatory processes contributing to understanding relevant physiological and pathological mechanisms
Catalog No. 50-000-64178 Supplier Apexbio Technology LLC Supplier No. B79570.05
8-Aza-ATP is a structural analog of ATP characterized by the substitution of the carbon atom at the 8-position of the purine ring with nitrogen resulting in an 8-azapurine nucleotide moiety Due to this structural alteration 8-Aza-ATP competitively interacts with ATP-binding sites on enzymes thereby modulating their catalytic activities This compound serves as a tool in biochemical research to elucidate enzyme-substrate recognition nucleotide binding specificity and catalytic mechanisms of ATP-dependent enzymes Additionally it is utilized in the study of cellular energy metabolism signal transduction pathways and nucleotide-mediated regulatory processes contributing to understanding relevant physiological and pathological mechanisms