Learn More
Medchemexpress LLC 8-Azaadenine | 1123-54-2 | 99.9% | 136.12 | 250 MG
SDP
Catalog No. 5000398562
Encompass_Preferred
8-Azaadenine is an inhibitor of Xanthine Oxidase with an IC50 of 0.54 μM and a Ki of 0.66 μM. It is produced via the metabolism of 8-Azaadenosine and can be incorporated into polynucleotide chains alongside 8-Azaguanine to impact purine metabolism. This product is used in cancer studies and can also be synthesized into various derivatives through alkylation reactions, which demonstrate antiviral activity.
  • Inhibits Xanthine Oxidase with an IC50 of 0.54 μM and a Kᵢ of 0.66 μM
  • Produced via the metabolism of 8-Azaadenosine
  • Incorporated into polynucleotide chains to affect purine metabolism
  • Used in the study of cancer
  • Can be synthesized into derivatives with antiviral activity
Catalog No. 50-003-98562 Supplier Medchemexpress LLC Supplier No. 250MGHYW008550
8-Azaadenine is an inhibitor of Xanthine Oxidase with an IC50 of 0.54 μM and a Ki of 0.66 μM. It is produced via the metabolism of 8-Azaadenosine and can be incorporated into polynucleotide chains alongside 8-Azaguanine to impact purine metabolism. This product is used in cancer studies and can also be synthesized into various derivatives through alkylation reactions, which demonstrate antiviral activity.
  • Inhibits Xanthine Oxidase with an IC50 of 0.54 μM and a Kᵢ of 0.66 μM
  • Produced via the metabolism of 8-Azaadenosine
  • Incorporated into polynucleotide chains to affect purine metabolism
  • Used in the study of cancer
  • Can be synthesized into derivatives with antiviral activity