Learn More
Cayman Chemical 8-AzaadenosIn 10mg
SDP
Catalog No. 502950260
Encompass_Preferred
A nucleoside analog and an ADAR1 inhibitor; inhibits RNA A-to-I editing in, as well as the invasion and migration of, TPC-1 and Cal-62 thyroid cancer cells at 1 and 2 µM; increases the ex vivo secretion of IFN-γ and TNF-α from CD8+ T cells isolated from mouse spleen in a model of HBV infection at 2 mg/kg; decreases serum HBsAg levels in the same model
Catalog No. 50-295-0260 Supplier Cayman Chemical Supplier No. 3462210 MG
A nucleoside analog and an ADAR1 inhibitor; inhibits RNA A-to-I editing in, as well as the invasion and migration of, TPC-1 and Cal-62 thyroid cancer cells at 1 and 2 µM; increases the ex vivo secretion of IFN-γ and TNF-α from CD8+ T cells isolated from mouse spleen in a model of HBV infection at 2 mg/kg; decreases serum HBsAg levels in the same model