Learn More
Medchemexpress LLC HY-115686 5mg , 8-Azaadenosine CAS:10299-44-2 Purity:>98%
SDP
Catalog No. 502259462
Encompass_Preferred
HY-115686 5mg 8-Azaadenosine CAS: 10299-44-2 8-Azaadenosine is a potent ADAR1 inhibitor and an A-to-I editing inhibitor. 8-Azaadenosine blocks RNA editing and inhibits proliferation, 3D growth, invasion, and migration in thyroid cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9462 Supplier Medchemexpress LLC Supplier No. HY1156865MG
HY-115686 5mg 8-Azaadenosine CAS: 10299-44-2 8-Azaadenosine is a potent ADAR1 inhibitor and an A-to-I editing inhibitor. 8-Azaadenosine blocks RNA editing and inhibits proliferation, 3D growth, invasion, and migration in thyroid cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.