Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-Azabicyclo[3.2.1]octan-3-ol, 8-methyl-, (3-endo)- | 120-29-6 | 100 G
SDP

Catalog No. 5000418011
Encompass_Preferred

Tropine is a tropane alkaloid that serves as a crucial intermediate for synthesizing anticholinergics and bronchodilators such as Atropine and Scopolamine. It is naturally found in Solanaceae Plants and *Przewalskia tangutica Maximo*.

  • Important intermediate for synthesis of anticholinergics and bronchodilators
  • Suitable for research and analytical applications
  • Appearance: white to light yellow solid
  • Purity (GC): 99.26%

Catalog No. 50-004-18011 Supplier Medchemexpress LLC Supplier No. HYN7061100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tropine is a tropane alkaloid that serves as a crucial intermediate for synthesizing anticholinergics and bronchodilators such as Atropine and Scopolamine. It is naturally found in Solanaceae Plants and *Przewalskia tangutica Maximo*.

  • Important intermediate for synthesis of anticholinergics and bronchodilators
  • Suitable for research and analytical applications
  • Appearance: white to light yellow solid
  • Purity (GC): 99.26%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.