Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Azahypoxanthine 98.0+%, TCI America™
SDP

Catalog No. A05551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A05551G Supplier TCI America Supplier No. A05551G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2683-90-1
Molecular Formula C4H3N5O
Molecular Weight (g/mol) 137.10
MDL Number MFCD00005804
InChI Key OEEYCNOOAHGFHL-UHFFFAOYSA-N
Synonym 8-azahypoxanthine, azahypoxanthine, 3h-1,2,3 triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-1,2,3,4,6-pentaazaindene, 7h-1,2,3-triazolo 4,5-d pyrimidin-7-one, 1,4-dihydro, 7-hydroxy-v-triazolo d pyrimidine, 1h-1,2,3-triazolo 4,5-d pyrimidin-7-ol, v-triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-3h-1,2,3 triazolo 4,5-d pyrimidine
PubChem CID 75895
IUPAC Name 2H,3H,7H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one
SMILES O=C1N=CN=C2NNN=C12

Specifications

Quantity 1 g
Formula Weight 137.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Azahypoxanthine
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.