Learn More
Medchemexpress LLC 8-azidoadenosine 5'-triphosphate trisodium | 00-00-0 | 98.1% | 614.14 | C10H12N8Na3O13P3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY134320B5MG
Encompass_Preferred
8-Azido-ATP trisodium is a photoreactive adenosine triphosphate analog supplied as the trisodium salt in solution. It is used to photo-crosslink and label ATP-binding proteins and serves as an azide-containing reagent for click chemistry and protein-nucleotide interaction studies.
  • Photoreactive ATP analog for photo-crosslinking and protein identification.
  • Azide functionality enables copper-catalyzed and strain-promoted click chemistry.
  • Supplied as trisodium salt in a solution form suitable for direct use or dilution.
  • High reported purity, supporting reproducible experimental results.
  • Storage guidelines support frozen stock solutions for long-term stability.
Catalog No. 50-002-21823