Learn More
Medchemexpress LLC 8-azidoadenosine 5'-triphosphate trisodium | 00-00-0 | 98.1% | C10H12N8Na3O13P3 | 10MG
SDP
Catalog No. 5000260519
Encompass_Preferred
8-Azido-ATP trisodium is a photoreactive adenosine triphosphate analog used for photoaffinity labeling and identification of ATP-binding proteins and nucleotide-interacting enzymes. Supplied as the trisodium salt with high purity (98.1%), it is intended for research applications involving UV-activated crosslinking to probe protein-nucleotide interactions.
  • Photoreactive azido group enables UV-activated crosslinking to binding partners.
  • Suitable for identifying ATP-binding proteins and nucleotide-interacting enzymes.
  • Available as the water-soluble trisodium salt for aqueous assays.
  • High purity suitable for analytical and biological experiments.
  • Compatible with downstream identification techniques such as SDS-PAGE and mass spectrometry.
Catalog No. 50-002-60519 Supplier Medchemexpress LLC Supplier No. HY134320B10MG
8-Azido-ATP trisodium is a photoreactive adenosine triphosphate analog used for photoaffinity labeling and identification of ATP-binding proteins and nucleotide-interacting enzymes. Supplied as the trisodium salt with high purity (98.1%), it is intended for research applications involving UV-activated crosslinking to probe protein-nucleotide interactions.
  • Photoreactive azido group enables UV-activated crosslinking to binding partners.
  • Suitable for identifying ATP-binding proteins and nucleotide-interacting enzymes.
  • Available as the water-soluble trisodium salt for aqueous assays.
  • High purity suitable for analytical and biological experiments.
  • Compatible with downstream identification techniques such as SDS-PAGE and mass spectrometry.