Learn More
Medchemexpress LLC 8-benzyl-4-oxo-8-azabicyclo[3.2.1]oct-2-ene-6,7-dicarboxylic acid | 960201-81-4 | 99.1% | 329.35 g/mol | C18H19NO5 | 10 MG
SDP
Catalog No. 5000240686
Encompass_Preferred
SD-1008 is a small-molecule JAK inhibitor that reduces STAT3-dependent signaling by inhibiting tyrosyl phosphorylation of STAT3, JAK2, and Src. It is supplied as a high-purity research reagent (C18H19NO5, 329.35 g/mol) intended for biochemical and cellular studies of JAK/STAT pathway modulation.
  • Potent JAK/STAT pathway inhibitor.
  • Reported purity 99.1%.
  • Suitable for biochemical and cellular assays.
  • Compact pack sizes for small-scale research use.
  • Molecular formula C18H19NO5 and MW 329.35 g/mol.
Catalog No. 50-002-40686 Supplier Medchemexpress LLC Supplier No. HY10759510MG
SD-1008 is a small-molecule JAK inhibitor that reduces STAT3-dependent signaling by inhibiting tyrosyl phosphorylation of STAT3, JAK2, and Src. It is supplied as a high-purity research reagent (C18H19NO5, 329.35 g/mol) intended for biochemical and cellular studies of JAK/STAT pathway modulation.
  • Potent JAK/STAT pathway inhibitor.
  • Reported purity 99.1%.
  • Suitable for biochemical and cellular assays.
  • Compact pack sizes for small-scale research use.
  • Molecular formula C18H19NO5 and MW 329.35 g/mol.