Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-benzyl-4-oxo-8-azabicyclo[3.2.1]oct-2-ene-6,7-dicarboxylic acid | 960201-81-4 | 99.1% | 329.35 g/mol | C18H19NO5 | 5 MG
SDP

Catalog No. 5000241614
Encompass_Preferred

SD-1008 is a small-molecule JAK inhibitor (CAS 960201-81-4) used in research to block tyrosyl phosphorylation of STAT3, JAK2, and Src and to enhance apoptosis in sensitive cell lines. It is supplied as a high-purity, solid reagent for biochemical and cellular studies.

  • Potent inhibition of JAK/STAT3 signaling, reducing phosphorylation of STAT3, JAK2, and Src.
  • High purity suitable for biochemical and cellular assays.
  • Molecular weight 329.35 g/mol and formula C18H19NO5 for accurate dosing.
  • Light yellow solid appearance, convenient for handling and dissolution.
  • Multiple package sizes available to support assay development and screening.
  • Storage recommendations: protect from light, store under inert gas, and maintain cold storage for solutions.

Catalog No. 50-002-41614 Supplier Medchemexpress LLC Supplier No. HY1075955MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SD-1008 is a small-molecule JAK inhibitor (CAS 960201-81-4) used in research to block tyrosyl phosphorylation of STAT3, JAK2, and Src and to enhance apoptosis in sensitive cell lines. It is supplied as a high-purity, solid reagent for biochemical and cellular studies.

  • Potent inhibition of JAK/STAT3 signaling, reducing phosphorylation of STAT3, JAK2, and Src.
  • High purity suitable for biochemical and cellular assays.
  • Molecular weight 329.35 g/mol and formula C18H19NO5 for accurate dosing.
  • Light yellow solid appearance, convenient for handling and dissolution.
  • Multiple package sizes available to support assay development and screening.
  • Storage recommendations: protect from light, store under inert gas, and maintain cold storage for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.