Learn More
Gilson Company Inc 8"BRASS SIEVE PAN,HALF HEIGHT
Catalog No. NC2221076
Encompass_Preferred
8" All Brass Sieve Pan, Half Height8in (203mm) diameter Brass half-height Pan for ASTM E11 8in Test Sieves is placed at the bottom of a sieve stack to collect fines passing through the bottom sieve.Features:Brass Pan for fines collectionFor use with 8in (203mm) diameter ASTM test sieves
This item is not returnable. View return policy
Catalog No. NC2221076 Supplier Gilson Company Inc Supplier No. V8BHXPN
8" All Brass Sieve Pan, Half Height8in (203mm) diameter Brass half-height Pan for ASTM E11 8in Test Sieves is placed at the bottom of a sieve stack to collect fines passing through the bottom sieve.Features:Brass Pan for fines collectionFor use with 8in (203mm) diameter ASTM test sieves