Learn More
Medchemexpress LLC 8-Bromo-2'-deoxyadenosine | 14985-44-5 | ≥98.0% | 330.14 | 50 MG
SDP
Catalog No. 5000451234
Encompass_Preferred
8-Bromo-2'-deoxyadenosine is a purine nucleoside analog. These analogs possess broad antitumor activity, specifically targeting indolent lymphoid malignancies. Their anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis.
  • Purine nucleoside analog
  • Has broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-004-51234 Supplier Medchemexpress LLC Supplier No. HYW01305250MG
8-Bromo-2'-deoxyadenosine is a purine nucleoside analog. These analogs possess broad antitumor activity, specifically targeting indolent lymphoid malignancies. Their anticancer mechanisms involve inhibiting DNA synthesis and inducing apoptosis.
  • Purine nucleoside analog
  • Has broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis