Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Bromo-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals

Catalog No. AAJ6535703
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6535703 1 g
AAJ65357MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6535703 Supplier Thermo Scientific Chemicals Supplier No. J6535703
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14985-44-5
Molecular Formula C10H12BrN5O3
Molecular Weight (g/mol) 330.14
MDL Number MFCD01630970
InChI Key NJBIVXMQFIQOGE-DWFKHXNSNA-N
Synonym 8-bromo-2'-deoxyadenosine, 2r,3s,5r-5-6-amino-8-bromo-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, 2r,3s,5r-5-6-amino-8-bromopurin-9-yl-2-hydroxymethyl oxolan-3-ol, adenosine,8-bromo-2'-deoxy, 2'-deoxy-8-bromoadenosine, 8-b-2-da, 8-bromo-2?-deoxyadenosine, adenosine, 8-bromo-2'-deoxy, 8-bromo-2-deoxyadenosine
PubChem CID 96547
SMILES NC1=C2N=C(Br)N([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

Color White to Yellow
Quantity 1 g
Solubility Information Soluble in DMSO or DMF
IUPAC Name (2R,3S,5R)-5-(6-amino-8-bromopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Formula Weight 330.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 8-Bromo-2'-deoxyadenosine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.