Learn More
Medchemexpress LLC 8-bromo-4H-oxadiazolo[3,4-c]benzoxazin-1-one | 204326-43-2 | 99.6% | 269.05 g/mol | C9H5BrN2O3 | 50MG
SDP
Catalog No. 5000237590
Encompass_Preferred
NS-2028 is a selective soluble guanylyl cyclase (sGC) inhibitor used in biochemical and cellular research to probe sGC signaling. It inhibits basal sGC activity with an IC50 of about 30 nM and NO-stimulated activity with an IC50 of about 200 nM. The compound is supplied in solid form or as a DMSO solution to support diverse experimental workflows.
  • Highly selective soluble guanylyl cyclase inhibitor for pathway interrogation.
  • High potency: IC50 ≈ 30 nM (basal) and ≈ 200 nM (NO-stimulated).
  • Suitable for biochemical and cell-based assays.
  • Available as solid and as DMSO solution formats for flexible dosing.
  • Molecular weight 269.05 g/mol and formula C9H5BrN2O3 for reference.
  • High purity suitable for research applications.
Catalog No. 50-002-37590 Supplier Medchemexpress LLC Supplier No. HY1237950MG
NS-2028 is a selective soluble guanylyl cyclase (sGC) inhibitor used in biochemical and cellular research to probe sGC signaling. It inhibits basal sGC activity with an IC50 of about 30 nM and NO-stimulated activity with an IC50 of about 200 nM. The compound is supplied in solid form or as a DMSO solution to support diverse experimental workflows.
  • Highly selective soluble guanylyl cyclase inhibitor for pathway interrogation.
  • High potency: IC50 ≈ 30 nM (basal) and ≈ 200 nM (NO-stimulated).
  • Suitable for biochemical and cell-based assays.
  • Available as solid and as DMSO solution formats for flexible dosing.
  • Molecular weight 269.05 g/mol and formula C9H5BrN2O3 for reference.
  • High purity suitable for research applications.