Learn More
Medchemexpress LLC 8-bromo-4H-oxadiazolo[3,4-c]benzoxazin-1-one | 204326-43-2 | MFCD01530052 | 99.6% | 269.05 g/mol | C9H5BrN2O3 | 100MG
SDP
Catalog No. 5000237496
Encompass_Preferred
NS-2028 is a selective small-molecule inhibitor of soluble guanylyl cyclase (sGC) used in biochemical and pharmacological research to probe nitric oxide-sGC-cGMP signaling pathways. It inhibits basal and NO-stimulated sGC activity with reported IC50 values of ~30 nM and ~200 nM and is supplied as a powder and as a DMSO solution for laboratory use.
  • Highly selective sGC inhibitor with low-nanomolar potency.
  • Used to study nitric oxide-sGC-cGMP signaling.
  • High reported purity (99.58%).
  • Molecular weight 269.05 g/mol; formula C9H5BrN2O3.
  • Available in small laboratory quantities and as a 10 mM DMSO solution.
  • Stable when stored as recommended (powder at -20°C).
Catalog No. 50-002-37496 Supplier Medchemexpress LLC Supplier No. HY12379100MG
NS-2028 is a selective small-molecule inhibitor of soluble guanylyl cyclase (sGC) used in biochemical and pharmacological research to probe nitric oxide-sGC-cGMP signaling pathways. It inhibits basal and NO-stimulated sGC activity with reported IC50 values of ~30 nM and ~200 nM and is supplied as a powder and as a DMSO solution for laboratory use.
  • Highly selective sGC inhibitor with low-nanomolar potency.
  • Used to study nitric oxide-sGC-cGMP signaling.
  • High reported purity (99.58%).
  • Molecular weight 269.05 g/mol; formula C9H5BrN2O3.
  • Available in small laboratory quantities and as a 10 mM DMSO solution.
  • Stable when stored as recommended (powder at -20°C).