Learn More
Enzo Life Sciences 8-Bromo-cADP-ribose (100µg). CAS: 151898-26-9
SDP
Supplier:  Enzo Life Sciences BMLCA4170100
Encompass_Preferred
cADPR antagonist. Blocks the cADPR-induced release of Ca2+ from sea urchin egg homogenates at 1-3.5 µM. Purity: ≥95%. Solubility: Soluble in water (5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0395