Learn More
Selleck Chemical LLC 8-Bromo-cAMP 100mg 76939-46-3
SDP
Supplier:  Selleck Chemical LLC S7857100MG
Encompass_Preferred
8-bromo-cAMP is a cell permeable analog of cAMP that activates cyclic-AMP-dependent protein kinase with a Ka value of 0.05 ?M; and a PKA activator. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5348