Learn More
Enzo Life Sciences 8-Bromo-cAMP, (10mg), CAS Number: 76939-46-3
SDP
Supplier:  Enzo Life Sciences BMLCN1150010
Encompass_Preferred
PKA activator. A cell-permeable cAMP analog which is more resistant to phosphodiesterases than cAMP and which preferentially activates cAMP-dependent protein kinase (PKA). Has increased metabolic stability and membrane permeability comapred with cAMP. Formula: C10H10BrN5O6P. Na. MW: 430.1. CAS Number: 76939-46-3. Purity: ≥98% (HPLC). Appearance: White to grey powder. Solubility: Soluble in water (100mg/ml). Use/Stability: Store, as supplied, at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C
Catalog No. NC9768490