Learn More
Selleck Chemical LLC 8-Bromo-cAMP
SDP
Catalog No. 5000001447
Encompass_Preferred
8-bromo-cAMP (8-Br-Camp) is a cell permeable analog of cAMP that activates cyclic-AMP-dependent protein kinase with a Ka value of 0 05 M and a PKA activator
Catalog No. 50-000-01447 Supplier Selleck Chemical LLC Supplier No. S785710MM/1ML
8-bromo-cAMP (8-Br-Camp) is a cell permeable analog of cAMP that activates cyclic-AMP-dependent protein kinase with a Ka value of 0 05 M and a PKA activator