Learn More
Medchemexpress LLC 8-Bromo-cAMP sodium salt | 76939-46-3 | 99.9% | 430.08 | 100 MG
SDP
Catalog No. 5000364981
Encompass_Preferred
8-Bromo-cAMP sodium salt is a cyclic AMP analog and an activator of cyclic AMP-dependent protein kinase (PKA). It has shown anti-proliferative and apoptotic effects against various cancer cells.
  • Enhances reprogramming efficiency of human neonatal foreskin fibroblast (HFF1) cells.
  • Induces apoptosis in esophageal cancer cell line (Eca-109).
  • Induces decidualization of human endometrial stromal cells.
  • Reduces tumor growth in CT26 tumor mice.
Catalog No. 50-003-64981 Supplier Medchemexpress LLC Supplier No. 100MGHY12306
8-Bromo-cAMP sodium salt is a cyclic AMP analog and an activator of cyclic AMP-dependent protein kinase (PKA). It has shown anti-proliferative and apoptotic effects against various cancer cells.
  • Enhances reprogramming efficiency of human neonatal foreskin fibroblast (HFF1) cells.
  • Induces apoptosis in esophageal cancer cell line (Eca-109).
  • Induces decidualization of human endometrial stromal cells.
  • Reduces tumor growth in CT26 tumor mice.